Anti-SLITRK1 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NOVUNBP1-49541EA 542 EUR
NOVUNBP1-49541 NOVUNBP1-49541SS
Anti-SLITRK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLITRK1 (SLIT and NTRK-like family, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse