Anti-KLRB1 Mouse Monoclonal Antibody [clone: 14F1F11]

Lieferant: Novus Biologicals
NOVUNBP2-14845EA 602 EUR
NOVUNBP2-14845
Anti-KLRB1 Mouse Monoclonal Antibody [clone: 14F1F11]
Antikörper
Type: Primary
Antigen: KLRB1 (killer cell lectin-like receptor subfamily B, member 1)
Clonality: monoclonal
Clone: 14F1F11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR