Anti-TLR4 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MTS510]

Lieferant: Novus Biologicals
NOVUNBP2-24450EA 634 EUR
NOVUNBP2-24450
Anti-TLR4 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: MTS510]
Antikörper
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: monoclonal
Clone: MTS510
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR