Anti-IL17A Mouse Monoclonal Antibody [clone: 4k5F6]

Lieferant: Novus Biologicals
NOVUNBP2-24765EA 641 EUR
NOVUNBP2-24765
Anti-IL17A Mouse Monoclonal Antibody [clone: 4k5F6]
Antikörper
Type: Primary
Antigen: IL17A (interleukin 17A)
Clonality: monoclonal
Clone: 4k5F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR