Anti-HES5 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP2-27174EA 546 EUR
NOVUNBP2-27174
Anti-HES5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HES5 (hes family bHLH transcription factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR