Anti-AGR2 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP2-27393EA 632 EUR
NOVUNBP2-27393
Anti-AGR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AGR2 (anterior gradient 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR