Anti-BST2 Mouse Monoclonal Antibody [clone: 2E2]

Lieferant: Novus Biologicals
NOVUNBP2-29621SSEA 251 EUR
NOVUNBP2-29621SS NOVUNBP2-29621
Anti-BST2 Mouse Monoclonal Antibody [clone: 2E2]
Antikörper
Type: Primary
Antigen: BST2 (bone marrow stromal cell antigen 2)
Clonality: monoclonal
Clone: 200
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR