Anti-GLI3 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP2-29627EA 428 EUR
NOVUNBP2-29627 NOVUNBP2-29627SS
Anti-GLI3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GLI3 (GLI family zinc finger 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR