Anti-Avi-tag Rabbit Polyclonal Antibody

Lieferant: GenScript Biotech
GENSA00674-40EA 169 EUR
GENSA00674-40
Anti-Avi-tag Rabbit Polyclonal Antibody
Antikörper
Type: Tag
Antigen: Avi-tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR