Anti-c-Myc tag Mouse Monoclonal Antibody

Lieferant: GenScript Biotech
GENSA00704-100EA 295 EUR
GENSA00704-100
Anti-c-Myc tag Mouse Monoclonal Antibody
Antikörper
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR