Anti-GST Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: GenScript Biotech
GENSA00866-100EA 304 EUR
GENSA00866-100
Anti-GST Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Tag
Antigen: GST
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR