Anti-SERPINH1 Mouse Monoclonal Antibody [clone: M16.10A1]

Lieferant: Oncogene
386023-50EA 516 EUR
386023-50
Anti-SERPINH1 Mouse Monoclonal Antibody [clone: M16.10A1]
Antikörper
Type: Primary
Antigen: SERPINH1 (serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1))
Clonality: monoclonal
Clone: M16.10A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR