Anti-Hsp90 (16F1) Rat Monoclonal Antibody

Lieferant: Oncogene
386041-50EA 516 EUR
386041-50
Anti-Hsp90 (16F1) Rat Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Hsp90 (16F1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR