Anti-ITGB6 Mouse Monoclonal Antibody [clone: 442.5C4]

Lieferant: Oncogene
407317-100EA 516 EUR
407317-100
Anti-ITGB6 Mouse Monoclonal Antibody [clone: 442.5C4]
Antikörper
Type: Primary
Antigen: ITGB6 (integrin, beta 6)
Clonality: monoclonal
Clone: 442.5C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR