Anti-ICAM1 Mouse Monoclonal Antibody

Lieferant: Oncogene
CP53-100EA 544 EUR
CP53-100
Anti-ICAM1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ICAM1 (intercellular adhesion molecule 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR