Anti-8-Hydroxyguanosine Goat Polyclonal Antibody

Lieferant: Oncogene
DR1001-100EA 531 EUR
DR1001-100
Anti-8-Hydroxyguanosine Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: 8-Hydroxyguanosine (8-hydroxyguanosine)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR