Anti-p53 (Ab-1) (Pantropic) (PAb421) Mouse Monoclonal Antibody [clone: PAb421]

Lieferant: Oncogene
OP03-100EA 544 EUR
OP03-100
Anti-p53 (Ab-1) (Pantropic) (PAb421) Mouse Monoclonal Antibody [clone: PAb421]
Antikörper
Type: Primary
Antigen: p53 (Ab-1) (Pantropic) (PAb421)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR