Anti-mTOR/FRAP (Ab-2) (22C2) Mouse Monoclonal Antibody

Lieferant: Oncogene
OP97-100EA 544 EUR
OP97-100
Anti-mTOR/FRAP (Ab-2) (22C2) Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: mTOR/FRAP (Ab-2) (22C2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR