Anti-Cleaved Bid (Ab-1) Rabbit Polyclonal Antibody
Lieferant: Oncogene
PC645-10EA 736 EUR
PC645-10
Anti-Cleaved Bid (Ab-1) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Cleaved Bid (Ab-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse