Anti-CAT Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-463EA 467 EUR
ROCK100-401-463
Anti-CAT Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-Chloramphenicol-Acetyl Transferase (CAT) Reacts with Pan

Type: Primary
Antigen: CAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR