Anti-Biotin Goat Antibody (FITC (Fluorescein isothiocyanate))

Lieferant: Rockland Immunochemicals
ROCK200-1298EA 5560 EUR
ROCK200-1298
Anti-Biotin Goat Antibody (FITC (Fluorescein isothiocyanate))
Antikörper
Type: Primary
Antigen: Biotin
Clonality:
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR