Anti-IgG F(ab')2 Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK201-1104EA 498 EUR
ROCK201-1104
Anti-IgG F(ab')2 Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR