Anti-GFP Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Rockland Immunochemicals
ROCK600-100-215EA 1040 EUR
ROCK600-100-215
Anti-GFP Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Primary Goat Anti-GFP Min X Hu Ms and Rt Serum Proteins Reacts with Pan

Recommended Dilutions: Immunofluorescence: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Full length
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR