Anti-FITC Goat Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
ROCK600-109-096EA 1060 EUR
ROCK600-109-096
Anti-FITC Goat Polyclonal Antibody (Texas Red®)
Antikörper
Anti-Fluorescein Antibody Texas Red® Conjugated is suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: 1:500 - 1:2500; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR