Anti-COL1A1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
ROCK600-406-103EA 814 EUR
ROCK600-406-103
Anti-COL1A1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Anti-COLLAGEN Type I Antibody Biotin Conjugated is suitable for western blot, immunoprecipitatioin, Flow Cytometry, and immunochemistry. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:3000 - 1:6000; Western Blot: 1:3000 - 1:6000; Immunochemistry: 1:50 - 1:200; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: COL1A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Bovine, Rat, Mouse, Mammalian
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR