Anti-V5 Tag Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029914EA 470 EUR
USBI029914
Anti-V5 Tag Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR