Anti-EPO Mouse Monoclonal Antibody [clone: 511CT20.2.4]

Lieferant: United States Biological
USBI030948EA 587 EUR
USBI030948
Anti-EPO Mouse Monoclonal Antibody [clone: 511CT20.2.4]
Antikörper
  • Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-16000

Type: Primary
Antigen: EPO (erythropoietin)
Clonality: Monoclonal
Clone: 511CT20.2.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR