Anti-RPS6KB2 Mouse Monoclonal Antibody [clone: 164CT21.2.2]
Lieferant: United States Biological
USBI031227EA 587 EUR
USBI031227
Anti-RPS6KB2 Mouse Monoclonal Antibody [clone: 164CT21.2.2]
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:1000-2000; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: Monoclonal
Clone: 164CT21.2.2
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse