Anti-Neurofilament H Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI031398EA 473 EUR
USBI031398
Anti-Neurofilament H Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:10000

Type: Primary
Antigen: Neurofilament H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR