Anti-VIM Mouse Monoclonal Antibody [clone: 13B868]

Lieferant: United States Biological
USBI031442EA 473 EUR
USBI031442
Anti-VIM Mouse Monoclonal Antibody [clone: 13B868]
Antikörper
Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:10000

Type: Primary
Antigen: VIM (vimentin)
Clonality: Monoclonal
Clone: 13B868
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR