Anti-AFF4 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI123043EA 522 EUR
USBI123043
Anti-AFF4 Mouse Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: AFF4 (AF4/FMR2 family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR