Anti-DPP4 Mouse Monoclonal Antibody [clone: 1C5]

Lieferant: United States Biological
USBI124592EA 522 EUR
USBI124592
Anti-DPP4 Mouse Monoclonal Antibody [clone: 1C5]
Antikörper
Type: Primary
Antigen: DPP4 (dipeptidyl-peptidase 4)
Clonality: monoclonal
Clone: 1C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR