Anti-NDFIP2 Mouse Polyclonal Antibody

Lieferant: US Biological
USBI130190EA 522 EUR
USBI130190
Anti-NDFIP2 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NDFIP2 (Nedd4 family interacting protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR