Anti-WWP1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI135419EA 561 EUR
USBI135419
Anti-WWP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: WWP1 (WW domain containing E3 ubiquitin protein ligase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR