Anti-IL7 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBI168297EA 817 EUR
USBI168297
Anti-IL7 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000

Type: Primary
Antigen: IL7 (interleukin 7)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR