Anti-CD27 Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: LG.3A10]

Lieferant: United States Biological
USBIC2379-09KEA 799 EUR
USBIC2379-09K
Anti-CD27 Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: LG.3A10]
Antikörper
Type: Primary
Antigen: CD27 (CD27 molecule)
Clonality: Monoclonal
Clone: LG.3A10
Conjugation: FITC (Fluorescein)
Epitope:
Host: Hamster
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR