Anti-CDR2 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC2581-01BEA 668 EUR
USBIC2581-01B
Anti-CDR2 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 2?g/ml

Type: Primary
Antigen: CDR2 (cerebellar degeneration-related protein 2, 62kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR