Anti-Complement C5b Neo-epitope, Human Mouse Monoclonal Antibody [clone: 5F208]

Lieferant: United States Biological
USBIC7850-30AEA 581 EUR
USBIC7850-30A
Anti-Complement C5b Neo-epitope, Human Mouse Monoclonal Antibody [clone: 5F208]
Antikörper
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Complement C5b Neo-epitope, Human
Clonality: Monoclonal
Clone: 5F208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Goat, Human, Monkey, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR