Anti-Desmoplakin 1,2 Mouse monoclonal antibody [clone: 5F190]

Lieferant: United States Biological
USBID3221-92EA 714 EUR
USBID3221-92
Anti-Desmoplakin 1,2 Mouse monoclonal antibody [clone: 5F190]
Antikörper
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: Desmoplakin 1,2
Clonality: monoclonal
Clone: 5F190
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR