Anti-Dopamine Beta Hydroxylase Mouse Monoclonal Antibody [clone: 10B2322]

Lieferant: United States Biological
USBID9010-02BEA 837 EUR
USBID9010-02B
Anti-Dopamine Beta Hydroxylase Mouse Monoclonal Antibody [clone: 10B2322]
Antikörper
Type: Primary
Antigen: Dopamine Beta Hydroxylase
Clonality: monoclonal
Clone: 10B2322
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR