Anti-EPO Mouse Monoclonal Antibody [clone: 5E592]

Lieferant: United States Biological
USBIE3455-15EA 508 EUR
USBIE3455-15
Anti-EPO Mouse Monoclonal Antibody [clone: 5E592]
Antikörper
Type: Primary
Antigen: EPO (erythropoietin)
Clonality: monoclonal
Clone: 5E592
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR