Anti-GRK6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBIG8972-25C-APCEA 852 EUR
USBIG8972-25C-APC
Anti-GRK6 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: GRK6 (G protein-coupled receptor kinase 6)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR