Anti-ITM2B Goat Polyclonal Antibody

Lieferant: United States Biological
USBII7661-29BEA 551 EUR
USBII7661-29B
Anti-ITM2B Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:8000; Western Blot: 1-3g/ml

Type: Primary
Antigen: ITM2B (integral membrane protein 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR