Anti-KLK5 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIK0005-08GEA 551 EUR
USBIK0005-08G
Anti-KLK5 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:4000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: KLK5 (kallikrein-related peptidase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR