Anti-Matrix Metalloproteinase 14 Chicken Polyclonal Antibody

Lieferant: United States Biological
USBIM2428-99EA 691 EUR
USBIM2428-99
Anti-Matrix Metalloproteinase 14 Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Matrix Metalloproteinase 14
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Bovine, Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR