Anti-PRKCI Rabbit Polyclonal Antibody

Lieferant: US Biological
P9103-70L
USBIP9103-70LEA 649 EUR
USBIP9103-70L
Anti-PRKCI Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: PRKCI (protein kinase C, iota)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR