Anti-PRKG1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIP9103-72Z4EA 470 EUR
USBIP9103-72Z4
Anti-PRKG1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: PRKG1 (protein kinase, cGMP-dependent, type I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR