Anti-PTK2B Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIP9400-09EA 592 EUR
USBIP9400-09
Anti-PTK2B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTK2B (protein tyrosine kinase 2 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR