Anti-RAE1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIR0485-50CEA 551 EUR
USBIR0485-50C
Anti-RAE1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:64000; Western Blot: 0. 03-0.1 ?g/ml

Type: Primary
Antigen: RAE1 (ribonucleic acid export 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR