Anti-RCBTB2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIR1260-05BEA 551 EUR
USBIR1260-05B
Anti-RCBTB2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:2000

Type: Primary
Antigen: RCBTB2 (regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR